4-(propoxymethyl)phenylboronic acid 95%

Lieferant: Apollo Scientific

OR908461-1G
APOSOR908461-1GEA 451 EUR
APOSOR908461-1G
4-(propoxymethyl)phenylboronic acid 95%
4-(propoxymethyl)phenylboronic acid
Formel: C₁₀H₁₅BO₃
Molecular Weight: 194.035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14687314
CAS-Nummer: 160061-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR