(3-(Pyridin-3-yl)phenyl)boronic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-(3-Pyridyl)benzeneboronic acid, [3-(Pyridin-3-yl)phenyl]boronic acid

OR908505-5G
APOSOR908505-5GEA 132 EUR
APOSOR908505-5G
(3-(Pyridin-3-yl)phenyl)boronic acid 95%
(3-(Pyridin-3-yl)phenyl)boronic acid
Formel: C₁₁H₁₀BNO₂
Molecular Weight: 199.016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658918
CAS-Nummer: 351422-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR