1-Chloro-3-methyl-5-nitrobenzene 97%

Lieferant: Apollo Scientific

OR908513-25G
APOSOR908513-25GEA 0 EUR
APOSOR908513-25G
1-Chloro-3-methyl-5-nitrobenzene 97%
1-Chloro-3-methyl-5-nitrobenzene
Formel: C₇H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09955417
CAS-Nummer: 16582-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR