5-Methyl-2-(methylthio)phenylboronic acid 96%

Lieferant: Apollo Scientific

OR908552-1G
APOSOR908552-1GEA 451 EUR
APOSOR908552-1G
5-Methyl-2-(methylthio)phenylboronic acid 96%
5-Methyl-2-(methylthio)phenylboronic acid
Formel: C₈H₁₁BO₂S
Molecular Weight: 182.048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384185
CAS-Nummer: 1259443-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR