2-Cyclopropyloxybenzeneboronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

OR908570-1G OR908570-250MG
APOSOR908570-1GEA 302 EUR
APOSOR908570-1G APOSOR908570-250MG
2-Cyclopropyloxybenzeneboronic acid, pinacol ester
2-Cyclopropyloxybenzeneboronic acid, pinacol ester
Formel: C₁₅H₂₁BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16994462
CAS-Nummer: 1119090-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR