4-Methylpyrazole-5-boronic acid, pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
OR908586-250MG
OR908586-100MG
APOSOR908586-250MGEA
207
EUR
APOSOR908586-250MG
APOSOR908586-100MG
4-Methylpyrazole-5-boronic acid, pinacol ester
4-Methylpyrazole-5-boronic acid, pinacol ester
Formel:
C₁₀H₁₇BN₂O₂ Lagertemperatur: Gefrierschrank |
MDL:
MFCD18250651 CAS-Nummer: 1440520-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...