Prop-1-ynylboronic acid pinacol ester 96%

Lieferant: Apollo Scientific

OR908925-1G
APOSOR908925-1GEA 270 EUR
APOSOR908925-1G
Prop-1-ynylboronic acid pinacol ester 96%
Prop-1-ynylboronic acid pinacol ester
Formel: C₉H₁₅BO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13182123
CAS-Nummer: 347389-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR