6-Methyl-3-nitroimidazo[1,2-a]pyridin 98%

Lieferant: Apollo Scientific

OR908963-1G
APOSOR908963-1GEA 155 EUR
APOSOR908963-1G
6-Methyl-3-nitroimidazo[1,2-a]pyridin 98%
6-Methyl-3-nitroimidazo[1,2-a]pyridin
Formel: C₈H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06496196
CAS-Nummer: 67625-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR