5-Chloro[1,2,4]triazolo[4,3-a]pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
5-Chloro-[1,2,4]triazolo[4,3-a]pyridine
OR909064-1G
APOSOR909064-1GEA
79.9
EUR
APOSOR909064-1G
5-Chloro[1,2,4]triazolo[4,3-a]pyridine 95%
5-Chloro[1,2,4]triazolo[4,3-a]pyridine
Formel:
C₆H₄ClN₃ Molecular Weight: 153.569 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109431 CAS-Nummer: 27187-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...