2-Brom-4-iodtoluol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-4-iodo-1-methylbenzene, 1-Bromo-5-iodo-2-metnylbenzene

OR909079-100G
APOSOR909079-100GEA 165 EUR
APOSOR909079-100G
2-Brom-4-iodtoluol 98%
2-Brom-4-iodtoluol
Formel: C₇H₆BrI
Molecular Weight: 296.9300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779014
CAS-Nummer: 26670-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR