1-Boc-piperidin 95%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl Piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)piperidine, Piperidine-1-carboxylic acid tert-Butyl ester, tert-butyl piperidine-1-carboxylate, 1-Boc-piperidine

OR909204-100G
APOSOR909204-100GEA 95.7 EUR
APOSOR909204-100G
1-Boc-piperidin 95%
1-Boc-piperidin
Formel: C₁₀H₁₉NO₂
Molecular Weight: 185.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093935
CAS-Nummer: 75844-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR