4,5-Dichlor-o-toluidin 98%

Lieferant: Apollo Scientific

Synonyme: 4,5-Dichloro-2-methylaniline

OR909235-1G OR909235-5G
APOSOR909235-1GEA 74.1 EUR
APOSOR909235-1G APOSOR909235-5G
4,5-Dichlor-o-toluidin 98%
4,5-Dichlor-o-toluidin
Formel: C₇H₇Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD11855825
CAS-Nummer: 2387-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR