1-(4-Methoxyphenyl)cyclopentancarbonsäure 97%
Lieferant: Apollo Scientific
OR909246-1G
APOSOR909246-1GEA
197
EUR
APOSOR909246-1G
1-(4-Methoxyphenyl)cyclopentancarbonsäure 97%
1-(4-Methoxyphenyl)cyclopentanecarboxylic acid
Formel:
C₁₃H₁₆O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001374 CAS-Nummer: 43050-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...