1-(2-Chlorophenyl)cyclopropanecarboxylic acid 97%
Lieferant: Apollo Scientific
OR909253-1G
OR909253-5G
APOSOR909253-1GEA
24.4
EUR
APOSOR909253-1G
APOSOR909253-5G
1-(2-Chlorophenyl)cyclopropanecarboxylic acid 97%
Formel:
C₁₀H₉ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07374435 CAS-Nummer: 122143-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...