4-methyl-1,3-benzothiazole-2-thiol 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methylbenzo[d]thiazole-2(3H)-thione

OR909266-5G
APOSOR909266-5GEA 69.6 EUR
APOSOR909266-5G
4-methyl-1,3-benzothiazole-2-thiol 95%
4-methyl-1,3-benzothiazole-2-thiol
Formel: C₈H₇NS₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11044826
CAS-Nummer: 2268-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR