1,4,5,6-Tetrahydrocyclopenta[c]pyrazole hydrochloride 98%

Lieferant: Apollo Scientific

OR909280-1G
APOSOR909280-1GEA 219 EUR
APOSOR909280-1G
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole hydrochloride 98%
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole hydrochloride
Formel: C₆H₈N₂·ClH
Molecular Weight: 144,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11983586
CAS-Nummer: 2214-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR