4-Bromo-3-isopropyl-1H-pyrazole 98%

Lieferant: Apollo Scientific

OR909286-5G
APOSOR909286-5GEA 221 EUR
APOSOR909286-5G
4-Bromo-3-isopropyl-1H-pyrazole 98%
4-Bromo-3-isopropyl-1H-pyrazole
Formel: C₆H₉BrN₂
Molecular Weight: 189.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16249535
CAS-Nummer: 60061-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR