5-Chloro-1,3-dihydro-2H-benzimidazol-2-one 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-1H-benzo[d]imidazol-2(3H)-one

OR909307-5G
APOSOR909307-5GEA 91.5 EUR
APOSOR909307-5G
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one 95%
5-Chloro-1,3-dihydro-2H-benzimidazol-2-one
Formel: C₇H₅ClN₂O
Molecular Weight: 168.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00493866
CAS-Nummer: 2034-23-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR