2-Methyl-4-nitroanisol 98%
Lieferant: Apollo Scientific
Synonyme:
1-Methoxy-2-methyl-4-nitrobenzol, Methyl-4-nitro-o-tolylether, 2-Methoxy-5-nitrotoluol
OR909315-100G
APOSOR909315-100GEA
82.8
EUR
APOSOR909315-100G
2-Methyl-4-nitroanisol 98%
2-Methyl-4-nitroanisol
Formel:
C₈H₉NO₃ Molecular Weight: 167.16 g/mol Siedepunkt: 311 Schmelzpunkt: 96 Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787583 CAS-Nummer: 50741-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...