N-Methyl-1H-pyrrole-2-carboxamide 97%

Lieferant: Apollo Scientific

OR909318-1G OR909318-5G
APOSOR909318-1GEA 145 EUR
APOSOR909318-1G APOSOR909318-5G
N-Methyl-1H-pyrrole-2-carboxamide 97%
N-Methyl-1H-pyrrole-2-carboxamide
Formel: C₆H₈N₂O
Molecular Weight: 124.141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143555
CAS-Nummer: 132911-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR