N-Methyl-1H-pyrrole-2-carboxamide 97%
Lieferant: Apollo Scientific
OR909318-1G
OR909318-5G
APOSOR909318-1GEA
145
EUR
APOSOR909318-1G
APOSOR909318-5G
N-Methyl-1H-pyrrole-2-carboxamide 97%
N-Methyl-1H-pyrrole-2-carboxamide
Formel:
C₆H₈N₂O Molecular Weight: 124.141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15143555 CAS-Nummer: 132911-42-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...