6-Chlor-7-methylchinoxalin 98%
Lieferant: Apollo Scientific
OR909336-1G
OR909336-25G
OR909336-5G
APOSOR909336-1GEA
145
EUR
APOSOR909336-1G
APOSOR909336-25G
APOSOR909336-5G
6-Chlor-7-methylchinoxalin 98%
6-Chloro-7-methylquinoxaline
Formel:
C₉H₇ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD15143565 CAS-Nummer: 646504-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...