1-(3-Chlorophenyl)pyrrolidin-2-one 98%
Lieferant: Apollo Scientific
OR909413-10G
OR909413-1G
OR909413-5G
APOSOR909413-10GEA
754
EUR
APOSOR909413-10G
APOSOR909413-1G
APOSOR909413-5G
1-(3-Chlorophenyl)pyrrolidin-2-one 98%
1-(3-Chlorophenyl)pyrrolidin-2-one
Formel:
C₁₀H₁₀ClNO Molecular Weight: 195.646 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00522616 CAS-Nummer: 24051-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...