1-Methyl-4-(hydroxyethyl)piperidine 98%

Lieferant: Apollo Scientific

Synonyme: 2-(1-Methylpiperidino)ethanol, 2-(1-Methylpiperidin-4-yl)ethanol

OR909465-1G
APOSOR909465-1GEA 223 EUR
APOSOR909465-1G
1-Methyl-4-(hydroxyethyl)piperidine 98%
1-Methyl-4-(hydroxyethyl)piperidine
Formel: C₈H₁₇NO
Molecular Weight: 143.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13188565
CAS-Nummer: 21156-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR