2-Chloro-4-piperidin-1-ylpyrimidine 98%
Lieferant: Apollo Scientific
OR909497-1G
APOSOR909497-1GEA
258
EUR
APOSOR909497-1G
2-Chloro-4-piperidin-1-ylpyrimidine 98%
2-Chloro-4-piperidin-1-ylpyrimidine
Formel:
C₉H₁₂ClN₃ Molecular Weight: 197,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01912571 CAS-Nummer: 5429-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...