4-(Cyclopentyloxy)benzylamine 98%

Lieferant: Apollo Scientific

OR909511-1G
APOSOR909511-1GEA 531 EUR
APOSOR909511-1G
4-(Cyclopentyloxy)benzylamine 98%
4-(Cyclopentyloxy)benzylamine
Formel: C₁₂H₁₇NO
Molecular Weight: 191,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09733523
CAS-Nummer: 864266-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR