2-Methoxy-4-pyrrolidinopyrimidine 97%

Lieferant: Apollo Scientific

OR909525-1G OR909525-5G
APOSOR909525-1GEA 219 EUR
APOSOR909525-1G APOSOR909525-5G
2-Methoxy-4-pyrrolidinopyrimidine 97%
2-Methoxy-4-pyrrolidinopyrimidine
Formel: C₉H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD22383693
CAS-Nummer: 1393442-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR