2-Methoxy-4-morpholinopyrimidine 98%

Lieferant: Apollo Scientific

OR909557-1G OR909557-5G
APOSOR909557-1GEA 219 EUR
APOSOR909557-1G APOSOR909557-5G
2-Methoxy-4-morpholinopyrimidine 98%
2-Methoxy-4-morpholinopyrimidine
Formel: C₉H₁₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22421507
CAS-Nummer: 1400644-32-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR