N-(3-Cyanophenyl)-2,5-dimethylpyrrole 98%

Lieferant: Apollo Scientific

OR909573-1G OR909573-25G OR909573-5G
APOSOR909573-1GEA 145 EUR
APOSOR909573-1G APOSOR909573-25G APOSOR909573-5G
N-(3-Cyanophenyl)-2,5-dimethylpyrrole 98%
N-(3-Cyanophenyl)-2,5-dimethylpyrrole
Formel: C₁₃H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09063237
CAS-Nummer: 204142-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR