4-Benzylmorpholine 97%

Lieferant: Apollo Scientific

OR909577-1G OR909577-5G
APOSOR909577-1GEA 0 EUR
APOSOR909577-1G APOSOR909577-5G
4-Benzylmorpholine 97%
4-Benzylmorpholine
Formel: C₁₁H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00090503
CAS-Nummer: 10316-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR