2-Butanoyl-3-picoline 98%

Lieferant: Apollo Scientific

OR909609-1G
APOSOR909609-1GEA 270 EUR
APOSOR909609-1G
2-Butanoyl-3-picoline 98%
2-Butanoyl-3-picoline
Formel: C₁₀H₁₃NO
Molecular Weight: 163,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16087793
CAS-Nummer: 1248102-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR