(1-Phenylcyclobutyl)methylamine hydrochloride 95%

Lieferant: Apollo Scientific

OR909629-1G
APOSOR909629-1GEA 270 EUR
APOSOR909629-1G
(1-Phenylcyclobutyl)methylamine hydrochloride 95%
(1-Phenylcyclobutyl)methylamine hydrochloride
Formel: C₁₁H₁₅N·ClH
Molecular Weight: 197,704 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864745
CAS-Nummer: 209907-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR