4,7-Dimethyl-benzothiazol-2-ylamine 96%

Lieferant: Apollo Scientific

OR909631-10G OR909631-1G OR909631-5G
APOSOR909631-10GEA 438 EUR
APOSOR909631-10G APOSOR909631-1G APOSOR909631-5G
4,7-Dimethyl-benzothiazol-2-ylamine 96%
4,7-Dimethyl-benzothiazol-2-ylamine
Formel: C₉H₁₀N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD04971844
CAS-Nummer: 78584-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR