3-Amino-3-(2-pyridinyl)acrylonitrile 98%

Lieferant: Apollo Scientific

OR909633-1G OR909633-5G
APOSOR909633-1GEA 0 EUR
APOSOR909633-1G APOSOR909633-5G
3-Amino-3-(2-pyridinyl)acrylonitrile 98%
3-Amino-3-(2-pyridinyl)acrylonitrile
Formel: C₈H₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01871647
CAS-Nummer: 55330-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR