Ethyl-5-cyclopropyl-1-methyl-1H-pyrazole-3-carboxylate 98%

Lieferant: Apollo Scientific

OR909675-1G OR909675-5G
APOSOR909675-1GEA 219 EUR
APOSOR909675-1G APOSOR909675-5G
Ethyl-5-cyclopropyl-1-methyl-1H-pyrazole-3-carboxylate 98%
Ethyl-5-cyclopropyl-1-methyl-1H-pyrazole-3-carboxylate
Formel: C₁₀H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD15072122
CAS-Nummer: 1223748-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR