1-(Piperidin-3-yl)ethanone hydrochloride 98%

Lieferant: Apollo Scientific

OR909708-1G OR909708-5G
APOSOR909708-1GEA 0 EUR
APOSOR909708-1G APOSOR909708-5G
1-(Piperidin-3-yl)ethanone hydrochloride 98%
1-(Piperidin-3-yl)ethanone hydrochloride
Formel: C₇H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD12022655
CAS-Nummer: 89895-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR