1-(Methoxymethyl)pyrrole-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR909724-1G
APOSOR909724-1GEA 364 EUR
APOSOR909724-1G
1-(Methoxymethyl)pyrrole-2-carboxylic acid 98%
1-(Methoxymethyl)pyrrole-2-carboxylic acid
Formel: C₇H₉NO₃
Molecular Weight: 155,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18810222
CAS-Nummer: 172805-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR