4-Methoxy-N-methylanilin 98%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-p-anisidine

OR909746-25G
APOSOR909746-25GEA 63.8 EUR
APOSOR909746-25G
4-Methoxy-N-methylanilin 98%
4-Methoxy-N-methylanilin
Formel: C₈H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00008399
CAS-Nummer: 5961-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR