2-Methoxy-3-nitrobenzonitrile 98%
Lieferant: Apollo Scientific
OR909773-1G
OR909773-250MG
OR909773-5G
APOSOR909773-1GEA
221
EUR
APOSOR909773-1G
APOSOR909773-250MG
APOSOR909773-5G
2-Methoxy-3-nitrobenzonitrile 98%
2-Methoxy-3-nitrobenzonitrile
Formel:
C₈H₆N₂O₃ Molecular Weight: 178.145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18398370 CAS-Nummer: 121001-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...