2-Methoxy-3-nitrobenzonitrile 98%

Lieferant: Apollo Scientific

OR909773-1G OR909773-250MG OR909773-5G
APOSOR909773-1GEA 221 EUR
APOSOR909773-1G APOSOR909773-250MG APOSOR909773-5G
2-Methoxy-3-nitrobenzonitrile 98%
2-Methoxy-3-nitrobenzonitrile
Formel: C₈H₆N₂O₃
Molecular Weight: 178.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398370
CAS-Nummer: 121001-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR