Methyl-1-methylpiperidine-4-carboxylate 97%
Lieferant: Apollo Scientific
Synonyme:
Methyl 1-methylpiperidine-4-carboxylate
OR909799-100G
APOSOR909799-100GEA
95.7
EUR
APOSOR909799-100G
Methyl-1-methylpiperidine-4-carboxylate 97%
Methyl-1-methylpiperidine-4-carboxylate
Formel:
C₈H₁₅NO₂ Molecular Weight: 157.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658483 CAS-Nummer: 1690-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...