3-(p-Tolyl)propionsäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Methylphenyl)propionsäure, 3-(4-Methylphenyl)propansäure

OR909967-100G
APOSOR909967-100GEA 206 EUR
APOSOR909967-100G
3-(p-Tolyl)propionsäure 97%
3-(p-Tolyl)propionsäure
Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016561
CAS-Nummer: 1505-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR