5-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-[1,2,4]triazolo[1,5-a]pyridine
OR910006-1G
APOSOR910006-1GEA
63.8
EUR
APOSOR910006-1G
5-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
5-Bromo[1,2,4]triazolo[1,5-a]pyridine
Formel:
C₆H₄BrN₃ Molecular Weight: 198,0201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699201 CAS-Nummer: 143329-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...