1-(Ethoxymethyl)imidazol 98%

Lieferant: Apollo Scientific

OR910009-1G
APOSOR910009-1GEA 270 EUR
APOSOR910009-1G
1-(Ethoxymethyl)imidazol 98%
1-(Ethoxymethyl)imidazole
Formel: C₆H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09878401
CAS-Nummer: 67319-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR