4-Bromo-1,5-dimethylimidazole 95%
Lieferant: Apollo Scientific
OR910018-1G
APOSOR910018-1GEA
723
EUR
APOSOR910018-1G
4-Bromo-1,5-dimethylimidazole 95%
4-Bromo-1,5-dimethylimidazole
Formel:
C₅H₇BrN₂ Molecular Weight: 175,026 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033089 CAS-Nummer: 158585-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...