1-(4-Nitrophenyl)-1H-pyrazole 98%
Lieferant: Apollo Scientific
OR910032-25G
APOSOR910032-25GEA
98.6
EUR
APOSOR910032-25G
1-(4-Nitrophenyl)-1H-pyrazole 98%
1-(4-Nitrophenyl)-1H-pyrazole
Formel:
C₉H₇N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00187828 CAS-Nummer: 3463-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...