3-Methyl-1-phenyl-1H-pyrazol 95%

Lieferant: Apollo Scientific

OR910108-5G
APOSOR910108-5GEA 81.2 EUR
APOSOR910108-5G
3-Methyl-1-phenyl-1H-pyrazol 95%
3-Methyl-1-phenyl-1H-pyrazole
Formel: C₁₀H₁₀N₂
Molecular Weight: 158.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003123
CAS-Nummer: 1128-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR