7-Bromo-1H-benzo[d][1,2,3]triazole 95%

Lieferant: Apollo Scientific

OR910218-1G
APOSOR910218-1GEA 0 EUR
APOSOR910218-1G
7-Bromo-1H-benzo[d][1,2,3]triazole 95%
7-Bromo-1H-benzo[d][1,2,3]triazole
Formel: C₆H₄BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11226749
CAS-Nummer: 1064721-11-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR