1-tert-Butoxycarbonyl-1,2,4-triazole 98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 1H-1,2,4-triazole-1-carboxylate, 1-Boc-1,2,4-triazole

OR910221-25G OR910221-5G
APOSOR910221-25GEA 316 EUR
APOSOR910221-25G APOSOR910221-5G
1-tert-Butoxycarbonyl-1,2,4-triazole 98%
1-tert-Butoxycarbonyl-1,2,4-triazole
Formel: C₇H₁₁N₃O₂
Molecular Weight: 169.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042665
CAS-Nummer: 41864-24-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR