5-Bromo-2-chlorothiazole 97%

Lieferant: Apollo Scientific

OR910270-1G OR910270-250MG OR910270-5G
APOSOR910270-1GEA 105 EUR
APOSOR910270-1G APOSOR910270-250MG APOSOR910270-5G
5-Bromo-2-chlorothiazole 97%
5-Bromo-2-chlorothiazole
Formel: C₃HBrClNS
Molecular Weight: 198.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702782
CAS-Nummer: 3034-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR